Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (47mm)
Product Code. 11828482
Change view
Click to view available options
Volume (Metric):
300 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11828482 300 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11828482 Supplier DWK Life Sciences Supplier No. 9537550000
  • ONLINE EXCLUSIVE PRICE
    3349.50 NOK
    2953.27 NOK / Pack of 1
    Save 396.23 NOK
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-06-2027
The KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (47mm) is a 47mm vacuum microfiltration assembly for general filtration, handling up to 500mL of sample liquids for analysis of particulate or microbiological contamination.
  • Supplied with a 40-60 micron porosity fritted glass support base, a 300mL graduated funnel, an anodized aluminum clamp and a #8 silicone stopper
  • Funnels are graduated from 100 to 250mL in 25mL increments
  • Prefilter size is 35mm
  • Approximate filter area is 9.6cm2
  • Connection to 1 liter filtration flask (available as an accessory) is made with a #8 silicone stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Disposable Reusable
Material Glass
Sterility Non-sterile
Includes Fritted glass support base, 300mL graduated funnel, anodized aluminum clamp and a #8 silicone stopper
Volume (Metric) 300 mL
Width (English) 3 in.
Width (Metric) 7.6 cm
Height (English) 9 in.
Height (Metric) 22.9 cm
For Use With (Equipment) Filtration Assembly and vacuum pump