Learn More
DWK Life Sciences KIMBLE™ KONTES™ Squibb Pear-Shaped Separatory Funnel with Standard Taper Stopper and PTFE Stopcock
Description
- Supplied with a standard taper ground glass stopper and a fully autoclavable PTFE stopcock plug
- Pear-shaped design for efficient liquid separation
- Lower stems have an inside diameter large enough so that a column of liquid will break with the stopcock closed, giving a more complete separation and eliminating the necessity of emptying the funnel
- Designed from ASTM Specification E1096, Type IV requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I requirements
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E1096, Type IV |
| Length (Metric) Stem | 60 mm |
| Quantity | 1 Ea. |
| Stopcock | 4; PTFE |
| Stopper Material | Glass |
| Product Type | Squibb Separatory Funnel |