Learn More
DWK Life Sciences Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve
Click to view available options
Overall Length (English):
4.72 in.
5.9 in.
6.49 in.
Width (English):
2.55 in.
2.75 in.
2.95 in.
3.14 in.
Height (English):
3.93 in.
4.72 in.
5.9 in.
Unit Size:
Each
Description
- Medium wall borosilicate glass
Specifications
Specifications
| Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
| Material | Borosilicate Glass |
| Diameter (English) Outer | 1.49 in. |
| Diameter (Metric) Outer | 3.8 cm |
| Overall Length (English) | 5.9 in. |
| Overall Length (Metric) | 15 cm |
| For Use With | 216050 or 216060 manifolds |
| Width (Metric) | 8 cm |
| Width (English) | 3.14 in. |
| Height (Metric) | 15 cm |
| Show More |