Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement PTFE/Silicone Septa
Description
- Excellent for use with open-top caps to access container contents with a syringe
- Highly chemically resistant PTFE facing maintains integrity of contents
- Silicone rubber backing allows repeated puncturing through the seal
- Resists coring
- Autoclavable
- Recommended for use in all autosamplers using screw thread vials
Specifications
Specifications
| Closure Size | For Thread Cap Size 24 |
| Product Type | Septa |
| Material | Silicone Rubber, PTFE |
| For Use With (Application) | Highly chemically resistant PTFE facing maintains integrity of contents, silicone rubber backing allows repeated puncturing through the seal |
| Thickness | 0.010 in. (PTFE), 0.090 in. (Silicon Rubber) |
| For Use With (Equipment) | Kontes™ K8976350800 and K897735130 Threaded NMR Tubes and 24 mm Threaded Open Top Cap, All autosamplers using screw thread vials |