Learn More
DWK Life Sciences™ Kimble™ Kontes™ NMR Polypropylene Sample Tube Rack Supports
Description
- 820-A wilmad equivalent
- Accommodates 72- 3 or 5mm tubes
Specifications
Specifications
| Holds | Tubes (3 or 5 mm) |
| Material | Polypropylene |
| Positions | 72 |
| Dimensions (L x W x H) | 8.37 x 4.5 x 8.75 in. (19.3 x 8.8 x 22.2cm) |
| Length (Metric) | 19.3 cm |
| Length (English) | 8.37 in. |
| Width (English) | 4.5 in. |
| Height (English) | 8.75 in. |
| Product Type | Tube Rack |
| Height (Metric) | 22.2 cm |
| Show More |