Learn More
DWK Life Sciences KIMBLE™ KONTES™ Hollow White Rubber Sleeve Stoppers
Product Code. 15857691
Change view
Click to view available options
Diameter (Metric):
1.4 cm
2.4 cm
Standard Taper Joint:
14/20
24/40; 24/20
Unit Size:
Each
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Standard Taper Joint Unit Size
15857691 1.4 cm 14/20 Pack of 50
12859518 1.4 cm 14/20 Each
12869518 2.4 cm 24/40; 24/20 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15857691 Supplier DWK Life Sciences Supplier No. 7742610014
  • ONLINE EXCLUSIVE PRICE
    546.70 NOK
    460.57 NOK / Pack of 50
    Save 86.13 NOK
    15% Off
 Restricted Item
Estimated Shipment: 07-12-2026
KIMBLE™ KONTES™ Hollow White Rubber Sleeve Stoppers fit securely into the neck of glassware apparatus. The plug-type design with sleeve extension provides a reliable seal, and the diaphragm can be punctured with a syringe needle. Rated for use up to 125°C for up to 8 hours.
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
  • Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
  • Diaphragm can be punctured with a syringe needle
  • Use for outgassing NMR sample tubes, as a pressure vent, or for adding reactants via a syringe
  • White rubber construction rated for use up to 125°C for up to 8 hours
  • Available in standard taper sizes to fit common laboratory glassware
Specifications
Diameter (Metric) 1.4 cm
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
Standard Taper Joint 14/20
Autoclavable Autoclavable
Color White
Diameter (Metric) Bottom 8 mm
Diameter (Metric) Outer 18 mm
Diameter (Metric) Top 14 mm
Material Rubber
Product Type Plug-type Sleeve Stopper
Shape Cylindrical
Sterility Non-sterile
Stopper Number 14