Learn More
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Product Code. 12859518
Change view
Click to view available options
Diameter (Metric):
1.4 cm
2.4 cm
Standard Taper Joint:
14/20
24/40; 24/20
Unit Size:
Each
Pack of 50
Product Code. Diameter (Metric) Standard Taper Joint unitSize
12859518 1.4 cm 14/20 Each
15857691 1.4 cm 14/20 Pack of 50
12869518 2.4 cm 24/40; 24/20 Pack of 50
3 options
Product Code. 12859518
Brand: DWK Life Sciences 7742610014
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
  • Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
  • Diaphragm can be punctured with a syringe needle
  • Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 1.4 cm
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
Standard Taper Joint 14/20
Color White
Material Rubber
Product Type Plug-type Sleeve Stopper