Learn More
DWK Life Sciences Kimble™ Flex-Column Chromatography Fittings: Adapter with 1/4-28 MaleThread to Male Luer
Product Code. 11892913 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With:
Flex-Column
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11892913 Flex-Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11892913 Supplier DWK Life Sciences Supplier No. 4204072113
  • ONLINE EXCLUSIVE PRICE
    148.50 NOK
    108.89 NOK / Pack of 1
    Save 39.61 NOK
    26% Off
 Restricted Item
Estimated Shipment: 08-03-2027
This ETFE FLEX-COLUMN™ fitting is a male adapter with 1/4-28 thread to male Luer.
  • ETFE FLEX-COLUMN™ Fitting
  • Male Luer to Male 1/4 in.-28 Thread
  • Case of 1
Specifications
Type Adapter
Thread Size 0.25-28
Port Type Male luer to male
For Use With Flex-Column
Material ETFE