Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Polyethylene Disc
Product Code. 11608228
Change view
Click to view available options
Type:
Disc 30 to 50 μm
Disc, 20 μm
Unit Size:
Pack of 100
Product Code. Type unitSize
11608228 Disc 30 to 50 μm Pack of 100
10749175 Disc, 20 μm Pack of 100
2 options
Product Code. 11608228
Brand: DWK Life Sciences 4201620000
Only null left
Add to basket
Add to basket
For Disposaflex Chromatography Columns
  • Sold 100/pack.
TRUSTED_SUSTAINABILITY
Specifications
Type Disc 30 to 50 μm
For Use With Disposaflex Chromatography Column
Material Polyethylene