Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Packing Reservoir, with Joints
Product Code. 13491239
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
13491239 100 mL Each
13511990 250 mL Each
2 options
Product Code. 13491239
Brand: DWK Life Sciences 4205700124
Only null left
Add to basket
Add to basket
These heavy duty reservoirs are designed for use with 420510 and 420550 series chromatography columns with and without fritted glass supports or in combination with 420575 series flow control adapters.
  • Reservoirs are available in a variety of sizes and joint configurations
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Type Packing Reservoir with Joints
Volume (Metric) 100 mL