Description
Generate over three times the torque of conventional stirring bars with Thermo Scientific™ Komet Stir Bars, significantly enhancing stirrer performance and applications versatility.
- High-energy Samarium-Cobalt (SmCo) magnetic core
- Superior mixing action at high speeds, and with viscous media
- Evenly coated PTFE surface prevents residue deposits
- Sterilize with steam to 249.8°F (121°C)
- Longlife stir bars are round with wear-resistant glide ring
Ordering Information:
- Available in assorted sizes
Specifications
Specifications
| For Use With (Equipment) | Stirrer Controller |
| Includes | Glide Ring |
| Coating | PTFE |
| Diameter (Metric) | 31 mm |
| Length (Metric) | 90 mm |
| Material | PTFE |
| Quantity | 1 Ea. |
| Shape | Sphere |
| Type | Stir Bar |