Learn More
Koflo Tube Mixer
Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
178 mm
190 mm
292 mm
302 mm
Unit Size:
Each
Description
- Able to mix light viscosity to high viscosity components.
- Accept standard tube fittings.
- These mixers consist of a series of fixed right- and left-hand elements.
- Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies.
- Mixers have plain ends and accept standard tube fittings.
Specifications
Specifications
| Diameter (English) | 0.51 in. |
| Diameter (Metric) | 13 mm |
| Length (English) | 11.9 in. |
| Length (Metric) | 302 mm |
| Material | Stainless Steel (316) |
| Type | Tube Mixer |
| For Use With (Application) | Mixing high-viscosity fluids |