missing translation for 'onlineSavingsMsg'
Learn More

KN-92 hydrochloride, MedChemExpress

Product Code. 16416598
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16416598 10 mg 10mg
16460246 2 mg 2mg
16442687 50 mg 50mg
16419167 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16416598 Supplier Medchem Express Supplier No. HY15517B
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KN-92 hydrochloride is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 hydrochloride is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93.

KN-92 hydrochloride is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 hydrochloride is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93.

Specifications

Chemical Name or Material KN-92 hydrochloride
CAS 1431698-47-3
Color White
Purity Grade Notes Research
Molecular Formula C24H26Cl2N2O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (101.33 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=CC=CC=C2CN(C/C=C/C3=CC=C(Cl)C=C3)C)=O.[H]Cl
Molecular Weight (g/mol) 493.45
Formula Weight 493.45
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.