Learn More
KLKB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27991-1-AP-20UL
 View more versions of this product
Product Code. 16845945
  • 1710.33 NOK / 20µL
Description
The antigen of this antibody just cover the heavy chain of KLKB1, so this antibody cannot recognize the light chain.
Plasma prekallikrein is a glycoprotein that participates in the surface-dependent activation of blood coagulation, fibrinolysis, kinin generation and inflammation. It is synthesized in the liver and secreted into the blood as a single polypeptide chain. Plasma prekallikrein is converted to plasma kallikrein by factor XIIa by the cleavage of an internal Arg-Ile bond. Plasma kallikrein therefore is composed of a heavy chain and a light chain held together by a disulphide bond. The heavy chain originates from the amino-terminal end of the zymogen and contains 4 tandem repeats of 90 or 91 amino acids. Each repeat harbors a novel structure called the apple domain. The heavy chain is required for the surface-dependent pro-coagulant activity of plasma kallikrein. The light chain contains the active site or catalytic domain of the enzyme and is homologous to the trypsin family of serine proteases. Plasma prekallikrein deficiency causes a prolonged activated partial thromboplastin time in patients.
Specifications
Specifications
KLKB1
Polyclonal
Unconjugated
KLKB1
Fletcher factor, Kininogenin, KLK3, KLKB1, Plasma kallikrein, Plasma prekallikrein, PPK
Rabbit
Antigen Affinity Chromatography
RUO
16621, 25048, 3818
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P03952, P14272, P26262
KLKB1
KLKB1 Fusion Protein Ag27491
20 μL
Primary
Rat, Human, Mouse