Learn More
KLHL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21191-1-AP-20UL
 View more versions of this product
Product Code. 16850815
  • 1710.33 NOK / 20µL
Description
The protein encoded by TCP1 epsilon is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Mutations in TCP1 epsilon cause hereditary sensory and autonomic neuropathy with spastic paraplegia (HSNSP). Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 5 and 13.
Specifications
Specifications
KLHL4
Polyclonal
Unconjugated
KLHL4
DKELCHL, kelch like 4 (Drosophila), Kelch like protein 4, KHL4, KIAA1687, KLHL4
Rabbit
Antigen Affinity Chromatography
RUO
237010, 317196, 56062
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9C0H6
Klhl4
KLHL4 Fusion Protein Ag15529
20 μL
Primary
Human, Rat, Mouse