missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11880-1-AP-20UL
This item is not returnable.
View return policy
Description
Kruppel like factor 4 (KLF4) protein is encoded by the KLF4 gene located on chromosome 9 in humans. It is also known as gut-enriched Kruppel-like factor (GKLF) or epithelial zinc-finger protein (EZF) and belongs to a family of conserved Kruppel-like factor family of DNA binding zinc finger transcription factors. KLF4 exerts its functional relevance across varied cellular pathways. As one of the four Yamanaka factors, it plays a key role in reprogramming differentiated cells into pluripotent stem cells. Most studies have focused on its specific role in endothelial cell signaling, adipogenesis, differentiation, embryogenesis and cell cycle control. Localization plays an important role in the biological significance of this protein wherein nuclear localization is associated with its function as an oncogene and cytosolic localization with tumor suppression specific effect.Specifications
| KLF4 | |
| Polyclonal | |
| Unconjugated | |
| KLF4 | |
| EZF, GKLF, KLF4, Krueppel like factor 4, Kruppel like factor 4 (gut) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114505, 16600, 9314 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43474, Q60793 | |
| KLF4 | |
| KLF4 Fusion Protein Ag2480 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction