Description
The KLF2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KLF2. This antibody recognizes human, mouse antigen. The KLF2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | KLF2 |
| Applications | Western Blot, ELISA |
| Concentration | 1000 μg/mL |
| Gene Symbols | KLF2, Krueppel like factor 2, Kruppel like factor 2 (lung), LKLF, Lung krueppel like factor |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 10365 |
| Target Species | Human, Mouse |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Show More |