missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF13 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | KLF13 |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898434
|
Proteintech
18352-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888434
|
Proteintech
18352-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Anti-RANTES factor of Late Activated T Lymphocytes-1 (RFLAT-1) polyclonal antiserum. RFLAT-1 is a novel protein belonging to the TFIIIA-like zinc finger protein superfamily and binds to the A site of the RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) chemokine. RANTES is involved in the generation of inflammatory infiltrate and HIV entry into immune cells.Specifications
| KLF13 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Human | |
| Q9Y2Y9 | |
| 499171, 51621 | |
| KLF13 Fusion Protein Ag13200 | |
| Primary | |
| -20°C | |
| KLF13 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BTE binding protein 3, BTEB3, FKLF2, KLF13, Krueppel like factor 13, Kruppel like factor 13, NSLP1, RFLAT 1, RFLAT1, Transcription factor BTEB3, Transcription factor NSLP1 | |
| KLF13 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title