Learn More
Ecolab™ Klercide™ 70/30 Denatured Ethanol Blended With WFI
Product Code. 15396068
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15396068 5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15396068 Supplier Ecolab™ Supplier No. 3078340
  • ONLINE EXCLUSIVE PRICE
    10610.25 NOK
    10134.25 NOK / Pack of 4
    Save 476.00 NOK
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
70% denatured ethanol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
  • The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
Specifications
Quantity 4 Pk.
Volume (Metric) 5 L