Learn More
KleenGuard™ A40 Light Duty Overshoe
Product Code. 12675576
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12675576 Universal Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12675576 Supplier KleenGuard™ Supplier No. 98700
  • ONLINE EXCLUSIVE PRICE
    4347.00 NOK
    2182.13 NOK / Pack of 200
    Save 2164.87 NOK
    49% Off
View all promotions
Estimated Shipment: 04-12-2026
Ideal for keeping footwear clean and covered. KleenGuard™ A40 Light Duty Overshoe reduces contamination in food processing and light industrial areas.
  • Single-use light duty overboots
  • Ideal for keeping footwear clean and covered
  • Reduce contamination in food processing and light industrial areas
  • PPE Category I - CE Simple
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Application) In industry and laboratories.
Material Polyblend
Size Universal
Toe Material Multi-layer Non-woven Barrier Laminate
Disposable Single-use
Gender Unisex
Waterproof Water Resistant
Product Type Overshoe