Learn More
Kimberly-Clark Professional™ Kleenex™ 8478 Standard Size 2 Ply Toilet Roll, 48 Rolls
Product Code. 17294243
Change view
Click to view available options
Dimensions (L x W):
12.4 x 10.4 cm
Unit Size:
48 rolls
Product Code. Dimensions (L x W) unitSize
17294243 12.4 x 10.4 cm 48 rolls
1 options
Product Code. 17294243
Brand: Kimberly-Clark Professional™ 8478
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Standard size 2 ply toilet paper rolls are made from 100% pure fibres, designed to create an enhanced washroom experience. The soft and luxurious toilet tissue sheets provide elevated care and home-like comfort for your workplace. 48 rolls x 200 white, 2 ply sheets (9,600 sheets)
  • Each individual toilet roll contains 200 white, 2 ply toilet paper sheets
  • High quality standard Kleenex™ toilet roll that creates an enhanced washroom experience
  • 2 ply toilet paper is soft and luxurious and made from 100% pure fibres to provide elevated care
  • For environmental assurance, toilet paper rolls are FSC certified and have reliable flushability
  • Compatible with a variety of Kimberly Clark toilet tissue dispenser options
TRUSTED_SUSTAINABILITY
Specifications
Product Type Toilet Paper Roll
Length (English) 23.6 in.
Length (Metric) 60 cm
Width (English) 15.7 in.
Width (Metric) 40 cm
Material Paper
Certifications/Compliance FSC certified
Color White
Dimensions (L x W) 12.4 x 10.4 cm
Format Roll
For Use With (Application) High Traffic Washrooms
Packaging Quantity 200 Sheets/Roll
Ply 2-ply
Size Small