Learn More
Kimberly-Clark™ Scott™ Essential™ Foam Everyday Hand Cleanser
Product Code. 12611376
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12611376 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12611376 Supplier Kimberly-Clark™ Supplier No. 6340
  • ONLINE EXCLUSIVE PRICE
    1575.00 NOK
    501.93 NOK / Pack of 6
    Save 1073.07 NOK
    68% Off
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Dispenses the ideal amount of soap for each wash, preventing overuse. Scott™ Essential™ Foam Everyday Hand Cleanser delivers up to twice as many washes per liter as conventional liquid soaps.
  • Pink foam soap with a luxury feel and subtle fragrance.
  • Supplied in strong, hygienically sealed cassettes that are quick and easy to install.
  • Provides up to 2,500 shots of soap.
  • Shrinks to save space when empty and are recyclable (when the pump has been removed).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Cleanser
Color Pink
Dimensions (L x W x H) 19 x 9 x 9 cm
For Use With (Application) Ideal for busy washrooms; cleaning hands and maintaining good hygiene
Quantity 6 Pk.
Size 6 x 1 L
Volume (English) 33.8 oz.
Volume (Metric) 1 L
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.