missing translation for 'onlineSavingsMsg'
Learn More

KLC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17668-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847604

  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
Specifications

Specifications

KLC2
Polyclonal
Unconjugated
KLC2
kinesin light chain 2, KLC 2, KLC2
Rabbit
Antigen Affinity Chromatography
RUO
16594, 309159, 64837
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88448, Q9H0B6
KLC2
KLC2 Fusion Protein Ag11158
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers