Learn More
SCHOTT AG Lighting and Imaging™ KL2500 LED Lightsource with Gooseneck Lightguide and EU Plug Set
Product Code. 17374323 Shop All Schott Products
Change view
Click to view available options
Bulb Type:
Multiple LEDs
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bulb Type Unit Size
17374323 Multiple LEDs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17374323 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 250400+400051+154202
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 13-01-2027
Set consisting of the KL2500 LED lightsource, 2-arm fiber optic gooseneck lightguide, and EU power cord
  • 1100 Lm lightflux
  • Patented white light engine
  • Slim ergonomical design
  • Continuous dimming from 0-100%
  • Extra-fine dimming option, down to 0,1% steps
  • SUSB Coonectivity
  • Two-position filter slider
  • Low-noise fan
  • Wide range power supply
  • Gooseneck light guide with eco-friendly Puravis™ fiber optics
  • Precise positioning offers complete control of light placement
  • Gooseneck branches are positioned in opposite directions to create near shadow-free illumination
  • Re-adjust with ease
Specifications
Type Lighting Set including Lightsource, Lightguide and Powercord
Bulb Type Multiple LEDs
Certifications/Compliance CE Certified, B EMC Class
Diameter (Metric) Gooseneck Lightguide Active Fiber Bundle 4.5 mm
Dimensions (L x W x H) 114 x 231 x 137 mm
For Use With (Application) Stereo Microscopy Illumination
Frequency 47/63 HZ
Includes Lightsource, Power suppy, Power cord and Lightguide
Lamp Life 50,000 Hours
Length (Metric) Gooseneck Lightguide 600 mm / Power Cord 1.8 m
Lighting Type LEDs