missing translation for 'onlineSavingsMsg'
Learn More

KKL-10, MedChemExpress

Product Code. p-200096562
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476347

Brand: Medchem Express HY101865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KKL-10 is a small-molecule ribosome rescue inhibitor with broad-spectrum antimicrobial activity against bacteria.

KKL-10 is a small-molecule ribosome rescue inhibitor with broad-spectrum antimicrobial activity against bacteria.

Specifications

Chemical Name or Material KKL-10
Physical Form Solid
CAS 952849-76-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C14H10BrN3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 2.78 mg/mL (7.63 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(Br)S1)NC2=NN=C(C3=CC=C(C)C=C3)O2
Molecular Weight (g/mol) 364.22
Formula Weight 364.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.