Learn More
Velp™ KjTabs VW - Kjeldahl Catalyst Tablets, Velp™
Product Code. 15571775 Shop All Velp Products
Change view
Click to view available options
Quantity:
1000/Pk., 1000 Pc./Box
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15571775 1000/Pk., 1000 Pc./Box Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15571775 Supplier Velp™ Supplier No. A00000282
 Restricted Item
Estimated Shipment: 11-12-2026
KjTabsTM VW Kjeldahl Catalyst Tablets are ideal for applications based on the Wieninger method. The perfect solutions to speed up the digestion of your samples.
  • The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
  • Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with catalyst tablets.
  • The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
  • KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
  • VTCT for tougher samples & even faster digestion (a scaled down version of VCT) 3.5g K2SO4+ 0.105g CuSO4x 5 H2O + 0.105g TiO21000 pcs
  • Most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
Chemical Name or Material VW
Color White
Quantity 1000/Pk., 1000 Pc./Box
Packaging Metal Tin
Grade Reagent
compliance-icons
Product Identifier
  • KJ tabs VW
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: copper sulphate pentahydrate, selenium