Description
- The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
- Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with catalyst tablets.
- The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
- KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
- VTCT for tougher samples & even faster digestion (a scaled down version of VCT) 3.5g K2SO4+ 0.105g CuSO4x 5 H2O + 0.105g TiO21000 pcs
- Most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
Specifications
| Chemical Name or Material | VTCT |
| Color | White |
| Quantity | 1000/Pk., 1000 Pc./Box |
| Packaging | Metal Tin |
| Grade | Reagent |