Learn More
Velp™ KjTabs VST - Kjeldahl Catalyst Tablets, Velp™
Product Code. 15533824 Shop All Velp Products
Change view
Click to view available options
Quantity:
1000/Pk., 1000 Pc./Box
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15533824 1000/Pk., 1000 Pc./Box Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15533824 Supplier Velp™ Supplier No. A00000277
 Restricted Item
Estimated Shipment: 16-12-2026
KjTabsTM VST Kjeldahl Catalyst Tablets are ideal for oily and fatty samples. The perfect solutions to speed up the digestion process.
  • The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
  • Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with catalyst tablets.
  • The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
  • KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
  • VTCT for tougher samples & even faster digestion (a scaled down version of VCT) 3.5g K2SO4+ 0.105g CuSO4x 5 H2O + 0.105g TiO21000 pcs
  • Most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
Chemical Name or Material VST
Color White
Quantity 1000/Pk., 1000 Pc./Box
Packaging Metal Tin
Grade Reagent