Learn More
Velp™ KjTabs VS - Kjeldahl Antifoam Tablets, Velp™
Product Code. 15581775 Shop All Velp Products
Change view
Click to view available options
Quantity:
1000/Pk., 1000 Pc./Box
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15581775 1000/Pk., 1000 Pc./Box Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15581775 Supplier Velp™ Supplier No. A00000283
 Restricted Item
Estimated Shipment: 14-12-2026
KjTabsTM VS Antifoam are perfect to reduce foam formation during the initial phase of the mineralization process.
  • Specific antifoaming tablets consisting of Potassium sulphate and Silicone.
  • Antifoaming agents are fundamental to the good outcome of the Kjeldahl analysis since the production of foam tends to reduce the original quantity of the sample during the initial phase of the mineralization process. This can occur in many agricultural samples or fat-rich products.
  • VS Antifoam used for foam reduction 0.97 g K2SO4 + 0.03 g Silicone, 1000 pcs.
  • Added in combination with KjTabs™ catalylst tablets.
  • Most suitable choice when performing Kjeldahl analysis in a great variety of industries.
Specifications
Chemical Name or Material VS
Color White
Quantity 1000/Pk., 1000 Pc./Box
Packaging Metal Tin
Grade Reagent