missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KiSS1R/GPR54 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1928
This item is not returnable.
View return policy
Description
KiSS1R/GPR54 Polyclonal antibody specifically detects KiSS1R/GPR54 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| KiSS1R/GPR54 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 3 - 5 μg/mL | |
| AXOR12hypogonadotropin-1, G protein-coupled receptor 54, GPR54kiSS-1R, G-protein coupled receptor 54, G-protein coupled receptor OT7T175, HOT7T175, Hypogonadotropin-1, kiSS-1 receptor, KISS1 receptor, KiSS-1R, Kisspeptins receptor, Metastin receptor | |
| Synthetic 20 amino acid peptide from C-terminus of human KiSS1R/GPR54. | |
| 0.05 mL | |
| GPCR | |
| 84634 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction