missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | KIR3DL3/CD158z |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CH21 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1 |
| Host Species | Mouse |
| Immunogen | Human KIR3DL3/CD158z transfectants |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?