Learn More
Lenz Laborglasintrumente Kipp Dispenser Heads
Click to view available options
Capacity (Metric):
1 mL
5 mL
50 mL
100 mL
Unit Size:
Each
Description
- Dispenser heads for Kipp dispensers
- With cone NS 29/32
- Made of DURAN™ tubing
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Glass |
| Standard Taper Joint | NS 29/32 |