Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats
Tienda Thermo Scientific ProductosDescripción
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Especificaciones
Especificaciones
| Format | 96 DW Comb |
| Capacity | 96 Samples |
| Content And Storage | KingFisher Apex 96 PCR tip comb (for Apex only) |
| Disposable | Single-use |
| For Use With (Equipment) | KingFisher Apex |
| Quantity | 50/Cs. |