Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats
Product Code. 12603126 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
KingFisher Apex
KingFisher Flex
KingFisher Flex and Presto
Quantity:
100/Cs.
48/Cs.
50/Cs.
80/Cs.
Unit Size:
Pack of 100
Pack of 48
Pack of 50
Pack of 80
Product Code. For Use With (Equipment) Quantity unitSize
12603126 KingFisher Flex 80/Cs. Pack of 80
16532452 KingFisher Apex 50/Cs. Pack of 50
10357939 KingFisher Flex and Presto 48/Cs. Pack of 48
12613126 KingFisher Flex 100/Cs. Pack of 100
17160069 KingFisher Apex 100/Cs. Pack of 100
17180069 KingFisher Apex 48/Cs. Pack of 48
6 options
Product Code. 12603126
Brand: Thermo Scientific™ 97002514
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Format 96-well Comb
Capacity 96 Samples
Description KingFisher 96 tip comb for KingFisher PCR magnets, 80 pcs (for Flex only)
For Use With (Equipment) KingFisher Flex
Quantity 80/Cs.