Learn More
Thermo Scientific™ KingFisher™ Flex 24 Deep Well head
Shop All Thermo Scientific ProductsDescription
The KingFisher™ Flex 24 Deep Well head a magnetic head for the KingFisherTM Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 24 deep-well format system. Both deep-well plates and KingFisher 96 plates can be used during the same run. Therefore, it is possible to start the processing by using larger volumes (in a deep-well plate) and elute the purified sample to a smaller volume (in a KingFisher 96 or MagMAX 96 standard plate).
Specifications
Specifications
| Format | 24 Deep-well Head |
| Capacity | 24 Samples |
| For Use With (Equipment) | KingFisher™ Flex |
| Product Line | KingFisher |
| Quantity | Each |
| Shipping Condition | Room Temperature |