Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Product Code. 11680243 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Barcoded Tip Comb
Combi Pack
Elution Strip
Elution Strip Cap
Tip Comb
For Use With (Equipment):
KingFisher Apex
KingFisher Duo Prime only
KingFisher Flex and Presto
Unit Size:
Each
Pack of 100
Pack of 40
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) unitSize
11680243 Combi Pack KingFisher Duo Prime only Each
16542452 Tip Comb KingFisher Apex Pack of 100
11744978 Tip Comb KingFisher Flex and Presto Pack of 100
12693116 Tip Comb KingFisher Duo Prime only Pack of 50
12633116 Elution Strip KingFisher Duo Prime only Pack of 40
15357317 Elution Strip Cap KingFisher Duo Prime only Pack of 40
17170069 Barcoded Tip Comb KingFisher Apex Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11680243 Supplier Thermo Scientific™ Supplier No. 97003530
Only null left
Add to basket
Edge
Add to basket
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Combi Pack
Format 96 DW Comb
Description KingFisher combi pack for microtiter 96 deep-well plate (includes 8 plates, 8 tip combs, and 8 elution strips and caps) (for Duo Prime only).
For Use With (Equipment) KingFisher Duo Prime only
Includes 8 plates, 8 tip combs, and 8 elution strips and caps
Product Line KingFisher
How many plates come with the KingFisher combi pack for microtiter 96 deep-well plate (Cat. No. 97003530)?
This kit is for the KingFisher Duo Prime only. It contains the following for processing 96 samples:
• 8 plates
• 8 tip combs
• 8 elution strips
• 8 elution strip caps
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
What is the difference between the KingFisher Duo and the KingFisher Duo Prime Purification Systems?
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
Can I reuse the Kingfisher Plastics for 96 deep-well format?
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
Are the KingFisher Plastics for 96 deep-well format CE marked?
The KingFisher Plastics for 96 deep-well format are not CE marked.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.