Learn More
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns Product Code.: 14851192
Estimated Shipment: 26-02-2026
  • ONLINE EXCLUSIVE PRICE
    3465.00 NOK
    2675.62 NOK / 1lb
    Save 789.38 NOK
    22% Off
Only null left
Add to basket
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Product Code. 14851192
6 mm, 1lb
Click to view available options
Diameter (Metric):
5 mm
6 mm
Unit Size:
1lb
Pack of 3000
Product Code. 14851192
Brand: DWK Life Sciences 135006
  • ONLINE EXCLUSIVE PRICE
    3465.00 NOK
    2675.62 NOK / 1lb
    Save 789.38 NOK
    22% Off
Only null left
Estimated Shipment: 26-02-2026
Only null left
Add to basket
Add to basket
Resist attack by liquids or vapors
May be used as mixing beads, boiling stones, or packing
Order Info
Packed in 1 lb. boxes
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Diameter (Metric) 6 mm
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns >