Learn More
DWK Life Sciences Kimble™ ShellVial with N-51A Borosilicate Glass Plain Top
Click to view available options
Volume (Metric):
4 mL
Unit Size:
Pack of 144
Description
- Heavy wall reduces breakage.
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 15 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 4 mL |
| Height (English) | 1.77 in. |
| Style | Plain top |
| Volume (English) | 1 dr. |
| Closure Material | Plastic |
| Closure Type | With Titeseal™ Open-bottom Plug Style Plastic Closures, Unattached |
| Height (Metric) | 45 mm |
| Show More |