missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure

Product Code. p-307150
Click to view available options
Capacity (Metric):
200 mL
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 15219383

Brand: DWK Life Sciences 61000G 200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Neutraglas™

  • No closure
  • GPI Neck Finish: 20A

TRUSTED_SUSTAINABILITY

Specifications

Capacity 6.76 oz., 200mL
Capacity (Metric) 200 mL
Capacity (English) 6.76 oz.
Diameter (English) 2.55 in.
Material Borosilicate Glass
Length (Metric) 114mm
Product Type Serum Bottle
Length (English) 4.48 in.
Closure Material Aluminum
Closure Size 20A
Diameter (Metric) 65 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.