Learn More
Kimble™ Low-form Griffin Beakers with Spout
Product Code. 11736333
Change view
Click to view available options
Capacity (Metric):
1500 mL
2000 mL
Graduation Range:
200 to 1400 mL
200 to 2000 mL
Increments:
100 mL
Unit Size:
Pack of 16
Pack of 4
Product Code. Capacity (Metric) Graduation Range Increments unitSize
11736333 2000 mL 200 to 2000 mL 100 mL Pack of 4
10798985 1500 mL 200 to 1400 mL 100 mL Pack of 16
2 options
Product Code. 11736333
Brand: Kimble™ 140002000
Only null left
Add to basket
Add to basket
Designed with a thick, slightly flared, beaded top, Kimble™ Low-form Griffin Beakers with Spout have excellent pouring characteristics and a uniform sidewall and bottom thickness design that renders them suitable for use on a hotplate.
  • Matte finish marking area for use with an ordinary pencil
  • Easy-to-read white graduated scale for measuring and/or mixing liquids
  • Design meets ASTM Specification E960, Type I requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±5%
Capacity (English) 67.62 oz.
Diameter (English) Outer 5.11 in.
Diameter (Metric) Outer 13 cm
Capacity (Metric) 2000 mL
Material Borosilicate Glass
For Use With (Application) For Measuring and Mixing Liquids
Graduation Range 200 to 2000 mL
Height (English) 7.48 in.
Height (Metric) 19 cm
Includes Spout
Increments 100 mL
Label Type Durable Matte Marking
Quantity 12 Pk.
Type Beaker
Graduated Yes
Certifications/Compliance ASTM E960
Scale Dual White Scale
Style Griffin