Learn More
DWK Life Sciences Kimble™ Kontes™ Erlenmeyer Flasks
Product Code. p-314050
Click to view available options
Capacity (Metric):
1000 mL
Height (Metric):
23 cm
Standard Taper Joint:
29/42
Unit Size:
Each
Product Code. 12857660
Brand: DWK Life Sciences 6170000729
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used to measure, mix, and store liquids
  • Single neck flask
  • Has standard taperouter joint
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (English) 9.05 in.
Height (Metric) 23 cm
Standard Taper Joint 29/42
Mouth Style Narrow