Learn More
DWK Life Sciences Kimble™ Kontes™ Erlenmeyer Flasks
Click to view available options
Capacity (Metric):
1000 mL
Height (Metric):
23 cm
Standard Taper Joint:
29/42
Unit Size:
Each
Description
Used to measure, mix, and store liquids
- Single neck flask
- Has standard taperouter joint
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Height (English) | 9.05 in. |
| Height (Metric) | 23 cm |
| Standard Taper Joint | 29/42 |
| Mouth Style | Narrow |