Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders with Safety-coated Tube
Product Code. 11858462
Change view
Click to view available options
Capacity:
0.1 oz., 3mL
0.16 oz., 5mL
0.59 oz., 15mL
Unit Size:
Each
Product Code. Capacity unitSize
11858462 0.1 oz., 3mL Each
11808512 0.59 oz., 15mL Each
11813153 0.16 oz., 5mL Each
3 options
Product Code. 11858462
Brand: DWK Life Sciences 8854600021
Only null left
Add to basket
Add to basket
Autoclavable transparent coating
  • With glass pestle
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.1 oz.
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
Autoclavable Autoclavable
Diameter (Metric) Shaft 6mm
Diameter (English) Shaft 0.23 in.
Capacity (Metric) 3 mL
Length (English) Shaft 8.14 in.
Length (English) Tube 4.72 in.
Length (Metric) Shaft 207mm
Length (Metric) Tube 120mm
Material Glass
Tube Diameter (English) Outside 0.62 in.
Tube Diameter (Metric) Outside 16mm
Capacity 0.1 oz., 3mL
Size 21