Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders with Safety-coated Tube
Product Code. 11808512
Click to view available options
Capacity:
0.1 oz., 3mL
0.16 oz., 5mL
0.59 oz., 15mL
Unit Size:
Each
Product Code. 11808512
Brand: DWK Life Sciences 8854600023
Only null left
Add to basket
Add to basket
Autoclavable transparent coating
  • With glass pestle
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.59 oz.
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
Autoclavable Autoclavable
Diameter (Metric) Shaft 10mm
Diameter (English) Shaft 0.39 in.
Capacity (Metric) 15 mL
Length (English) Shaft 10.9 in.
Length (English) Tube 6.88 in.
Length (Metric) Shaft 278mm
Length (Metric) Tube 175mm
Material Glass
Tube Diameter (English) Outside 0.98 in.
Tube Diameter (Metric) Outside 25mm
Capacity 0.59 oz., 15mL
Size 23