Learn More
DWK Life Sciences Kimble™ Kontes™ Closed System Sterile Tissue Grinder (Molded Tip)
Product Code. p-7237581
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacity:
0.5 oz., 15 mL
missing translation for 'unitSize'
Pack of 10
Product Code. Capacity unitSize
12643225 0.5 oz., 15 mL Pack of 10
1 missing translation for 'options'
Product Code. 12643225
missing translation for 'mfr': DWK Life Sciences 7496000015
Only null left
Add to basket
Add to basket
Includes
Fully assembled, 30% glass-filled pestles, sample tubes with caps, and adhesive labels.
Minimizes the risk of personal contact with samples
  • Tissues are ground within a sealed container
  • Narrow pestles eliminate the potential of sample overflow
  • Molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination
  • Gamma sterilized in easy-open HDPE packs
  • CS2 tissue grinders have a vitrified tip for more aggressive grinding of difficult samples
  • Polypropylene pestles and conical sample tubes with solid HDPE caps
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.5 oz.
Capacity (Metric) 15 mL
Includes Fully assembled, 30% Glass filled Polypropylene pestle, Polypropylene conical sample tube with solid HDPE cap and an adhesive label
Material Polypropylene
Capacity 0.5 oz., 15 mL