Learn More
DWK Life Sciences Kimble™ Headspace Vials
Description
- Finish: 20mm A/S
Order Info
100 vials/pack, 10 packs/case.
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 23 mm |
| Material | Silica Glass |
| Volume (Metric) | 20.0 mL |
| Height (English) | 2.95 in. |
| No. per Case | 1000 |
| Style | Beveled, Modified Top |
| Closure Type | Crimp Cap |
| Quantity | 1000 Cs. |
| Closure Size | 20 mm |
| Show More |