Learn More
DWK Life Sciences Wheaton™ Disposable Polystyrene Serological Pipets in Canister
Product Code. 16448955 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
Unit Size:
Pack of 500
Product Code. Volume (Metric) unitSize
16448955 1 mL Pack of 500
16458955 2 mL Pack of 500
16438955 10 mL Pack of 500
16468955 5 mL Pack of 500
4 options
Product Code. 16448955
Brand: DWK Life Sciences 5670011100
Only null left
Add to basket
Add to basket
For mass testing and serological work.
  • Calibrated to deliver to the tip
  • Features two rings near the top
  • Irradiation sterilized
  • Printed with chemically resistant organic black ink
TRUSTED_SUSTAINABILITY
Specifications
Aspirator Plug Yes
Material Plastic
Sterility Sterile
For Use With (Application) For tissue culture work, mass testing and serological work
Graduated Yes
Graduations 0.01 mL
Negative Graduations 0.3 mL
Quantity 500/Cs.
Tolerance ±0.02 mL
Packaging Type Canister
Volume (English) 0.03 oz.
Volume (Metric) 1 mL
Type Serological Pipet