Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Product Code. 11853133
Change view
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
24-400
Unit Size:
Pack of 144
Product Code. Closure Size unitSize
11853133 24-400 Pack of 144
17781078 18-400 Pack of 144
12184001 13-425 Pack of 144
15365536 15-425 Pack of 144
10701574 20-400 Pack of 144
5 options
Product Code. 11853133
Brand: DWK Life Sciences 7380224400
Only null left
Add to basket
Add to basket
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 24-400
Liner Material PTFE, Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Material Phenolic
Color Black
For Use With (Equipment) Sample Vials