Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Product Code. 11863033
Click to view available options
Color:
Black
Blue
White
Pre-slit:
No
Yes
Closure Size:
13-425
8-425
9-425
Unit Size:
Pack of 1000
Product Code. 11863033
Brand: DWK Life Sciences 73813BK8425
Only null left
Add to basket
Add to basket
Autoclavable and used for multiple injections
  • Manufactured using the highest quality raw materials
  • Designed for an ideal fit with Kimble Chase autosampler vials
  • Convenient assembly of caps and liners
  • Available with standard or pre-slit septa
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polypropylene
Pre-slit Yes
Insert Type Pre-Slit
Autoclavable Autoclavable
Closure Size 8-425