Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Click to view available options
Color:
Black
Blue
White
Pre-slit:
No
Yes
Closure Size:
13-425
8-425
9-425
Unit Size:
Pack of 1000
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Convenient assembly of caps and liners
- Available with standard or pre-slit septa
Specifications
Specifications
| Color | Black |
| Material | Polypropylene |
| Pre-slit | No |
| Autoclavable | Autoclavable |
| Closure Size | 8-425 |